Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAST1
Entrez Gene ID
22983
Gene Name
microtubule associated serine/threonine kinase 1
Gene Aliases
SAST, SAST170
Chromosome Location
Chr.19: 12833931 - 12874953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014975.2 NP_055790.1 19 - 20 2444 88
XM_011527805.2 XP_011526107.1 20 - 21 2341 88
XM_011527808.2 XP_011526110.1 8 - 9 1283 88
XM_017026501.1 XP_016881990.1 16 - 17 1897 88
GenBank mRNA AB023190.1 - 19 - 20 2355 88
BC027985.1 - 8 - 9 1162 88

View Full Product Details