The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003801.1 | NP_001003801.1 | 5 - 6 | 685 | 101 |
NM_001003802.1 | NP_001003802.1 | 6 - 7 | 674 | 101 | |
NM_003078.3 | NP_003069.2 | 6 - 7 | 713 | 101 | |
XM_011516521.2 | XP_011514823.1 | 5 - 6 | 792 | 101 | |
GenBank mRNA | AK127595.1 | - | 6 - 7 | 1221 | 101 |
AK297876.1 | - | 5 - 6 | 707 | 101 | |
AK309500.1 | - | 5 - 6 | 670 | 101 | |
AK316173.1 | - | 5 - 6 | 657 | 101 | |
AY450430.1 | - | 5 - 6 | 989 | 101 | |
AY450431.1 | - | 6 - 7 | 674 | 101 | |
BC002628.2 | - | 6 - 7 | 674 | 101 | |
BC110350.1 | - | 5 - 6 | 604 | 101 | |
BX648385.1 | - | 5 - 6 | 467 | 101 | |
U66619.1 | - | 6 - 7 | 722 | 101 |