The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017025012.1 | XP_016880501.1 | 5 - 6 | 595 | 75 |
GenBank mRNA | AK289715.1 | - | 5 - 6 | 605 | 75 |
BC057792.1 | - | 5 - 6 | 611 | 75 | |
BC069649.1 | - | 5 - 6 | 558 | 75 | |
BC074768.2 | - | 5 - 6 | 535 | 75 | |
CR541766.1 | - | 5 - 6 | 512 | 75 | |
M83670.1 | - | 5 - 6 | 559 | 75 |