Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM3A
Entrez Gene ID
60343
Gene Name
family with sequence similarity 3 member A
Gene Aliases
2.19, DLD, DXS560S, XAP-7
Chromosome Location
Chr.X: 154506159 - 154516249 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171132.2 NP_001164603.1 6 - 7 708 59
NM_001171133.2 NP_001164604.1 5 - 6 594 59
NM_001282311.1 NP_001269240.1 6 - 7 903 59
NM_001282312.1 NP_001269241.1 6 - 7 649 59
NM_021806.3 NP_068578.2 5 - 6 805 59
XM_005274716.3 XP_005274773.1 7 - 8 753 59
XM_017029700.1 XP_016885189.1 5 - 6 1025 59
XM_017029701.1 XP_016885190.1 6 - 7 928 59
XM_017029702.1 XP_016885191.1 5 - 6 541 59
GenBank mRNA AK057945.1 - 6 - 7 523 59
AK124890.1 - 2 - 3 2996 59
AK314682.1 - 5 - 6 723 59
BC002934.1 - 6 - 7 807 59
BC008912.2 - 5 - 6 456 59
BM457676.1 - 6 - 7 690 59
CX867071.1 - 5 - 6 512 59
X87193.1 - 5 - 6 612 59

View Full Product Details