Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD8
Entrez Gene ID
256471
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
CCMD, CLN7
Chromosome Location
Chr.4: 127917805 - 127965984 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152778.2 NP_689991.1 1 - 2 87 82
GenBank mRNA BC029503.1 - 1 - 2 52 82

View Full Product Details