Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARIH2
Entrez Gene ID
10425
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases
ARI2, TRIAD1
Chromosome Location
Chr.3: 48918820 - 48985541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317333.1 NP_001304262.1 13 - 14 1569 109
NM_001317334.1 NP_001304263.1 10 - 11 1031 109
NM_006321.3 NP_006312.1 12 - 13 1447 109
XM_011533265.1 XP_011531567.1 12 - 13 1504 109
XM_011533266.1 XP_011531568.1 13 - 14 1626 109
XM_011533267.1 XP_011531569.1 12 - 13 1558 109
XM_011533268.1 XP_011531570.1 11 - 12 1451 109
XM_011533270.1 XP_011531572.1 13 - 14 1538 109
XM_011533271.1 XP_011531573.1 12 - 13 1188 109
XM_011533273.1 XP_011531575.1 14 - 15 1569 109
XM_011533275.2 XP_011531577.1 12 - 13 1389 109
XM_011533276.1 XP_011531578.1 7 - 8 845 109
XM_017005531.1 XP_016861020.1 12 - 13 1489 109
XM_017005532.1 XP_016861021.1 11 - 12 1382 109
XM_017005533.1 XP_016861022.1 11 - 12 1898 109
XM_017005534.1 XP_016861023.1 11 - 12 1107 109
XM_017005535.1 XP_016861024.1 11 - 12 1315 109
XM_017005536.1 XP_016861025.1 14 - 15 1603 109
XM_017005537.1 XP_016861026.1 13 - 14 1469 109
XM_017005538.1 XP_016861027.1 13 - 14 1515 109
XM_017005539.1 XP_016861028.1 12 - 13 1119 109
XM_017005540.1 XP_016861029.1 14 - 15 1500 109
XM_017005541.1 XP_016861030.1 14 - 15 1570 109
GenBank mRNA AB208857.1 - 6 - 7 582 109
AF099149.1 - 11 - 12 1251 109
AF183427.1 - 11 - 12 1253 109
AJ130978.1 - 11 - 12 1251 109
AK091240.1 - 11 - 12 1994 109
AK223550.1 - 13 - 14 1553 109
AK307694.1 - 9 - 10 797 109
AK313434.1 - 10 - 11 1111 109
BC000422.2 - 12 - 13 1407 109
CR456785.1 - 10 - 11 1107 109

View Full Product Details