Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARIH2
- Entrez Gene ID
-
10425
- Gene Name
- ariadne RBR E3 ubiquitin protein ligase 2
- Gene Aliases
-
ARI2, TRIAD1
- Chromosome Location
-
Chr.3: 48918820 - 48985541 on Build GRCh38
View Full Product Details