Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARIH2
Entrez Gene ID
10425
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases
ARI2, TRIAD1
Chromosome Location
Chr.3: 48918820 - 48985541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317333.1 NP_001304262.1 6 - 7 850 82
NM_001317334.1 NP_001304263.1 3 - 4 312 82
NM_006321.3 NP_006312.1 5 - 6 728 82
XM_011533265.1 XP_011531567.1 5 - 6 716 82
XM_011533266.1 XP_011531568.1 6 - 7 838 82
XM_011533267.1 XP_011531569.1 5 - 6 770 82
XM_011533268.1 XP_011531570.1 4 - 5 663 82
XM_011533270.1 XP_011531572.1 6 - 7 750 82
XM_011533271.1 XP_011531573.1 5 - 6 400 82
XM_011533273.1 XP_011531575.1 7 - 8 781 82
XM_011533275.2 XP_011531577.1 5 - 6 670 82
XM_017005531.1 XP_016861020.1 5 - 6 770 82
XM_017005532.1 XP_016861021.1 4 - 5 663 82
XM_017005533.1 XP_016861022.1 4 - 5 1110 82
XM_017005534.1 XP_016861023.1 4 - 5 319 82
XM_017005535.1 XP_016861024.1 4 - 5 527 82
XM_017005536.1 XP_016861025.1 7 - 8 884 82
XM_017005537.1 XP_016861026.1 6 - 7 750 82
XM_017005538.1 XP_016861027.1 6 - 7 796 82
XM_017005539.1 XP_016861028.1 5 - 6 400 82
XM_017005540.1 XP_016861029.1 7 - 8 781 82
XM_017005541.1 XP_016861030.1 7 - 8 851 82
GenBank mRNA AF099149.1 - 4 - 5 532 82
AF183427.1 - 4 - 5 531 82
AJ130978.1 - 4 - 5 532 82
AK001800.1 - 3 - 4 412 82
AK223550.1 - 6 - 7 834 82
AK307694.1 - 2 - 3 124 82
AK313434.1 - 3 - 4 392 82
BC000422.2 - 5 - 6 688 82
BM928477.1 - 3 - 4 259 82
CR456785.1 - 3 - 4 388 82

View Full Product Details