Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
Chromosome Location
Chr.3: 129001604 - 129040742 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024768.2 NP_079044.2 6 - 7 1591 105
XM_005247781.3 XP_005247838.1 6 - 7 1619 105
XM_011513161.2 XP_011511463.1 6 - 7 1051 105
XM_011513164.2 XP_011511466.1 5 - 6 1664 105
GenBank mRNA AK022119.1 - 4 - 5 865 105
BC093800.1 - 2 - 3 302 105
BC112195.1 - 2 - 3 302 105

View Full Product Details