The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020133.2 | NP_064518.1 | 1 - 2 | 141 | 90 |
XM_005267053.3 | XP_005267110.1 | 1 - 2 | 127 | 90 | |
XM_006715514.3 | XP_006715577.1 | 1 - 2 | 127 | 90 | |
XM_017011059.1 | XP_016866548.1 | 1 - 2 | 174 | 90 | |
GenBank mRNA | AF156776.1 | - | 1 - 2 | 71 | 90 |
AJ420473.1 | - | 1 - 2 | 113 | 90 | |
AK074208.1 | - | 1 - 2 | 127 | 90 | |
AK295025.1 | - | 1 - 2 | 88 | 90 | |
AK295548.1 | - | 1 - 2 | 85 | 90 | |
AK295826.1 | - | 1 - 2 | 64 | 90 | |
AK299721.1 | - | 1 - 2 | 111 | 90 | |
AY358506.1 | - | 1 - 2 | 81 | 90 | |
BC020209.1 | - | 1 - 2 | 73 | 90 | |
BQ072907.1 | - | - | 141 | 90 |