The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031422.5 | NP_113610.2 | 5 - 6 | 538 | 86 |
XM_017026033.1 | XP_016881522.1 | 4 - 5 | 437 | 86 | |
GenBank mRNA | AF239821.1 | - | 5 - 6 | 415 | 86 |
AF332472.1 | - | 4 - 5 | 329 | 86 | |
AK093349.1 | - | 4 - 5 | 437 | 86 | |
AY358488.1 | - | 5 - 6 | 517 | 86 | |
BC025764.1 | - | 5 - 6 | 482 | 86 | |
KU178792.1 | - | 4 - 5 | 224 | 86 |