The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321833.1 | NP_001308762.1 | 17 - 18 | 2421 | 70 |
NM_001321834.1 | NP_001308763.1 | 17 - 18 | 3329 | 70 | |
NM_001321835.1 | NP_001308764.1 | 17 - 18 | 2064 | 70 | |
NM_001321836.1 | NP_001308765.1 | 18 - 19 | 2511 | 70 | |
NM_016207.3 | NP_057291.1 | 17 - 18 | 2192 | 70 | |
GenBank mRNA | AF171877.1 | - | 17 - 18 | 1970 | 70 |
AK025440.1 | - | 15 - 16 | 1851 | 70 | |
AK223487.1 | - | 17 - 18 | 1990 | 70 | |
AK298918.1 | - | 15 - 16 | 1785 | 70 | |
BC011654.2 | - | 17 - 18 | 2065 | 70 | |
BC014106.1 | - | 17 - 18 | 2982 | 70 | |
BC020211.1 | - | 17 - 18 | 1990 | 70 |