The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024513.3 | NP_078789.2 | 3 - 4 | 369 | 74 |
XM_006713333.3 | XP_006713396.1 | 4 - 5 | 1534 | 74 | |
XM_006713334.3 | XP_006713397.1 | 3 - 4 | 1450 | 74 | |
XM_011534111.2 | XP_011532413.1 | 4 - 5 | 447 | 74 | |
GenBank mRNA | AJ292348.2 | - | 3 - 4 | 353 | 74 |
AL832358.1 | - | 3 - 4 | 357 | 74 | |
BC101468.1 | - | 2 - 3 | 239 | 74 | |
BC101470.1 | - | 2 - 3 | 239 | 74 | |
BC143368.1 | - | 2 - 3 | 239 | 74 | |
BP249484.1 | - | 3 - 4 | 360 | 74 |