Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM30B
Entrez Gene ID
161291
Gene Name
transmembrane protein 30B
Gene Aliases
CDC50B
Chromosome Location
Chr.14: 61277370 - 61281812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017970.2 NP_001017970.1 1 - 1 1804 65
GenBank mRNA AK091169.1 - 1 - 1 1233 65
DA666826.1 - 1 - 1 123 65

View Full Product Details