The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014868.4 | NP_055683.3 | 12 - 13 | 2374 | 118 |
XM_005254013.2 | XP_005254070.1 | 13 - 14 | 2466 | 118 | |
XM_006719717.2 | XP_006719780.1 | 12 - 13 | 2409 | 118 | |
XM_017020282.1 | XP_016875771.1 | 13 - 14 | 2451 | 118 | |
XM_017020283.1 | XP_016875772.1 | 12 - 13 | 2409 | 118 | |
GenBank mRNA | AB027196.1 | - | 12 - 13 | 2338 | 118 |
AK094011.1 | - | 13 - 14 | 2275 | 118 | |
AK097807.1 | - | 13 - 14 | 2300 | 118 | |
AK126274.1 | - | 13 - 14 | 2179 | 118 | |
AK301608.1 | - | 13 - 14 | 2248 | 118 | |
AK315558.1 | - | 12 - 13 | 2356 | 118 | |
AL389976.1 | - | 11 - 12 | 1646 | 118 | |
BC016622.1 | - | 12 - 13 | 2338 | 118 | |
BC031596.1 | - | 12 - 13 | 2338 | 118 | |
BC101709.1 | - | 12 - 13 | 1964 | 118 | |
BC101715.1 | - | 12 - 13 | 1964 | 118 | |
BC143493.1 | - | 12 - 13 | 1963 | 118 | |
BT006974.1 | - | 12 - 13 | 1891 | 118 | |
D87451.2 | - | 12 - 13 | 1891 | 118 |