The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014868.4 | NP_055683.3 | 4 - 5 | 1130 | 87 |
XM_005254013.2 | XP_005254070.1 | 5 - 6 | 1207 | 87 | |
XM_006719717.2 | XP_006719780.1 | 4 - 5 | 1150 | 87 | |
XM_017020282.1 | XP_016875771.1 | 5 - 6 | 1207 | 87 | |
XM_017020283.1 | XP_016875772.1 | 4 - 5 | 1150 | 87 | |
GenBank mRNA | AB027196.1 | - | 4 - 5 | 1094 | 87 |
AK094011.1 | - | 5 - 6 | 1031 | 87 | |
AK097807.1 | - | 5 - 6 | 1056 | 87 | |
AK126274.1 | - | 5 - 6 | 935 | 87 | |
AK301608.1 | - | 5 - 6 | 1004 | 87 | |
AK315558.1 | - | 4 - 5 | 1112 | 87 | |
AL389976.1 | - | 3 - 4 | 387 | 87 | |
BC016622.1 | - | 4 - 5 | 1094 | 87 | |
BC031596.1 | - | 4 - 5 | 1094 | 87 | |
BC101709.1 | - | 4 - 5 | 720 | 87 | |
BC101715.1 | - | 4 - 5 | 720 | 87 | |
BC143493.1 | - | 4 - 5 | 719 | 87 | |
BT006974.1 | - | 4 - 5 | 647 | 87 | |
D87451.2 | - | 4 - 5 | 647 | 87 |