The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000152.4 | NP_000143.2 | 15 - 16 | 2583 | 145 |
NM_001079803.2 | NP_001073271.1 | 16 - 17 | 2398 | 145 | |
NM_001079804.2 | NP_001073272.1 | 15 - 16 | 2318 | 145 | |
XM_005257193.2 | XP_005257250.1 | 16 - 17 | 2384 | 145 | |
XM_005257194.4 | XP_005257251.1 | 16 - 17 | 2424 | 145 | |
GenBank mRNA | BC040431.1 | - | 16 - 17 | 2330 | 145 |
DQ907243.1 | - | 14 - 15 | 2186 | 145 | |
M34424.1 | - | 14 - 15 | 2218 | 145 | |
Y00839.1 | - | 15 - 16 | 2405 | 145 |