Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDC4
Entrez Gene ID
23644
Gene Name
enhancer of mRNA decapping 4
Gene Aliases
GE1, Ge-1, HEDL5, HEDLS, RCD-8, RCD8
Chromosome Location
Chr.16: 67873023 - 67884514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014329.4 NP_055144.3 3 - 4 593 122
GenBank mRNA AK128582.1 - 4 - 5 664 122
AK291049.1 - 3 - 4 361 122
AK302563.1 - 3 - 4 359 122
BC043616.1 - 3 - 4 496 122
BC064567.1 - 3 - 4 490 122
L26339.1 - 3 - 4 490 122

View Full Product Details