The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005914.2 | NP_001005914.1 | 13 - 14 | 1736 | 99 |
NM_001290060.1 | NP_001276989.1 | 12 - 13 | 1693 | 99 | |
NM_001290061.1 | NP_001276990.1 | 12 - 13 | 1542 | 99 | |
NM_001290062.1 | NP_001276991.1 | 6 - 7 | 959 | 99 | |
NM_001290063.1 | NP_001276992.1 | 6 - 7 | 833 | 99 | |
NM_004636.3 | NP_004627.1 | 13 - 14 | 1739 | 99 | |
NR_110697.1 | 4 - 5 | 618 | 99 | ||
GenBank mRNA | AB083186.1 | - | 12 - 13 | 1489 | 99 |
AB209322.1 | - | 11 - 12 | 1919 | 99 | |
AF217991.1 | - | 12 - 13 | 1502 | 99 | |
AK092182.1 | - | 4 - 5 | 580 | 99 | |
AK293678.1 | - | 6 - 7 | 792 | 99 | |
BC009113.1 | - | 9 - 10 | 1443 | 99 | |
BC013975.2 | - | 12 - 13 | 1516 | 99 | |
BC024220.2 | - | 12 - 13 | 1516 | 99 | |
BC044602.1 | - | 6 - 7 | 921 | 99 | |
U28369.1 | - | 12 - 13 | 1682 | 99 |