The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004668.2 | NP_004659.2 | 4 - 5 | 503 | 76 |
XM_006716168.2 | XP_006716231.1 | 4 - 5 | 561 | 76 | |
XM_011516670.2 | XP_011514972.1 | 4 - 5 | 569 | 76 | |
XM_011516671.2 | XP_011514973.1 | 5 - 6 | 636 | 76 | |
XM_011516672.2 | XP_011514974.1 | 5 - 6 | 622 | 76 | |
XM_011516673.2 | XP_011514975.1 | 4 - 5 | 524 | 76 | |
XM_011516674.2 | XP_011514976.1 | 4 - 5 | 561 | 76 | |
XM_017012772.1 | XP_016868261.1 | 5 - 6 | 541 | 76 | |
GenBank mRNA | AF016833.2 | - | 4 - 5 | 503 | 76 |
BC120872.1 | - | 4 - 5 | 475 | 76 |