The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000820.3 | NP_000811.1 | 3 - 4 | 466 | 91 |
GenBank mRNA | AK290803.1 | - | 3 - 4 | 355 | 91 |
BC038984.1 | - | 3 - 4 | 414 | 91 | |
BX380508.2 | - | 3 - 4 | 464 | 91 | |
L13720.1 | - | 3 - 4 | 417 | 91 |