Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBBP8
Entrez Gene ID
5932
Gene Name
RB binding protein 8, endonuclease
Gene Aliases
COM1, CTIP, JWDS, RIM, SAE2, SCKL2
Chromosome Location
Chr.18: 22914121 - 23026486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002894.2 NP_002885.1 13 - 14 2361 92
NM_203291.1 NP_976036.1 13 - 14 2331 92
NM_203292.1 NP_976037.1 13 - 14 2412 92
XM_005258325.2 XP_005258382.1 13 - 14 2292 92
XM_005258326.3 XP_005258383.1 12 - 13 2222 92
XM_006722519.2 XP_006722582.1 15 - 16 2433 92
XM_006722520.2 XP_006722583.1 14 - 15 2369 92
XM_006722521.2 XP_006722584.1 13 - 14 2429 92
XM_011526132.2 XP_011524434.1 14 - 15 2380 92
XM_017025915.1 XP_016881404.1 11 - 12 1911 92
XM_017025916.1 XP_016881405.1 14 - 15 2452 92
GenBank mRNA AF043431.1 - 13 - 14 2328 92
AK292481.1 - 13 - 14 2292 92
BC030590.1 - 13 - 14 2159 92
BX648221.1 - 13 - 14 2386 92
U72066.1 - 13 - 14 2329 92

View Full Product Details