Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
Chromosome Location
Chr.3: 96814581 - 97761532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173655.3 NP_775926.1 12 - 13 1358 134
GenBank mRNA AL133666.1 - 11 - 12 1305 134

View Full Product Details