The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244682.1 | NP_001231611.1 | 4 - 5 | 292 | 67 |
NM_014352.3 | NP_055167.2 | 4 - 5 | 402 | 67 | |
XM_011542739.2 | XP_011541041.1 | 6 - 7 | 617 | 67 | |
XM_011542740.2 | XP_011541042.1 | 5 - 6 | 475 | 67 | |
XM_011542741.2 | XP_011541043.1 | 3 - 4 | 326 | 67 | |
XM_011542742.2 | XP_011541044.1 | 5 - 6 | 580 | 67 | |
XM_011542743.2 | XP_011541045.1 | 3 - 4 | 2721 | 67 | |
XM_017017487.1 | XP_016872976.1 | 4 - 5 | 402 | 67 | |
GenBank mRNA | AF133895.1 | - | 4 - 5 | 259 | 67 |
AF162278.1 | - | 4 - 5 | 366 | 67 | |
AJ012214.1 | - | 3 - 4 | 252 | 67 | |
AK291993.1 | - | 4 - 5 | 402 | 67 | |
AK302212.1 | - | 4 - 5 | 292 | 67 | |
BC101649.1 | - | 4 - 5 | 337 | 67 | |
BC143593.1 | - | 5 - 6 | 384 | 67 | |
BC143594.1 | - | 4 - 5 | 337 | 67 | |
BC143595.1 | - | 4 - 5 | 335 | 67 |