The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000408.4 | NP_000399.3 | 13 - 14 | 1904 | 131 |
NM_001083112.2 | NP_001076581.2 | 13 - 14 | 2130 | 131 | |
XM_005246469.2 | XP_005246526.1 | 13 - 14 | 1843 | 131 | |
XM_011510977.2 | XP_011509279.1 | 13 - 14 | 1898 | 131 | |
XM_011510978.2 | XP_011509280.1 | 14 - 15 | 1948 | 131 | |
XM_011510980.2 | XP_011509282.1 | 8 - 9 | 1129 | 131 | |
XM_017003830.1 | XP_016859319.1 | 13 - 14 | 1984 | 131 | |
XM_017003831.1 | XP_016859320.1 | 12 - 13 | 1793 | 131 | |
GenBank mRNA | AB209399.1 | - | 13 - 14 | 1899 | 131 |
AF311325.1 | - | 14 - 15 | 1940 | 131 | |
AK093198.1 | - | 14 - 15 | 1939 | 131 | |
AK290057.1 | - | 13 - 14 | 2065 | 131 | |
AK291065.1 | - | 13 - 14 | 2143 | 131 | |
AK292817.1 | - | 13 - 14 | 1903 | 131 | |
AK299645.1 | - | 9 - 10 | 1245 | 131 | |
U12424.1 | - | 13 - 14 | 1893 | 131 | |
U36310.1 | - | 13 - 14 | 1873 | 131 |