The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000408.4 | NP_000399.3 | 15 - 16 | 2084 | 166 |
NM_001083112.2 | NP_001076581.2 | 15 - 16 | 2310 | 166 | |
XM_005246469.2 | XP_005246526.1 | 15 - 16 | 2023 | 166 | |
XM_011510977.2 | XP_011509279.1 | 15 - 16 | 2078 | 166 | |
XM_011510978.2 | XP_011509280.1 | 16 - 17 | 2128 | 166 | |
XM_011510980.2 | XP_011509282.1 | 10 - 11 | 1309 | 166 | |
XM_017003830.1 | XP_016859319.1 | 15 - 16 | 2164 | 166 | |
XM_017003831.1 | XP_016859320.1 | 14 - 15 | 1973 | 166 | |
GenBank mRNA | AB209399.1 | - | 15 - 16 | 2079 | 166 |
AF311325.1 | - | 16 - 17 | 2120 | 166 | |
AK093198.1 | - | 16 - 17 | 2119 | 166 | |
AK290057.1 | - | 15 - 16 | 2245 | 166 | |
AK291065.1 | - | 15 - 16 | 2323 | 166 | |
AK292817.1 | - | 15 - 16 | 2083 | 166 | |
AK299645.1 | - | 11 - 12 | 1425 | 166 | |
BC019874.1 | - | 10 - 11 | 1238 | 166 | |
U12424.1 | - | 15 - 16 | 2073 | 166 | |
U36310.1 | - | 15 - 16 | 2053 | 166 |