The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000729.5 | NP_000720.1 | 3 - 4 | 896 | 73 |
GenBank mRNA | AK289716.1 | - | 1 - 2 | 71 | 73 |
AK300784.1 | - | 1 - 2 | 71 | 73 | |
BC093055.1 | - | 1 - 2 | 71 | 73 | |
BC111026.1 | - | 3 - 4 | 871 | 73 | |
CB961869.1 | - | 1 - 2 | 98 | 73 | |
CD250282.1 | - | 1 - 2 | 231 | 73 |