The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032445.2 | NP_115821.2 | 11 - 12 | 1553 | 101 |
XM_017022670.1 | XP_016878159.1 | 10 - 11 | 1975 | 101 | |
XM_017022671.1 | XP_016878160.1 | 10 - 11 | 1973 | 101 | |
XM_017022672.1 | XP_016878161.1 | 10 - 11 | 1982 | 101 | |
XM_017022673.1 | XP_016878162.1 | 10 - 11 | 1976 | 101 | |
XM_017022674.1 | XP_016878163.1 | 5 - 6 | 1919 | 101 | |
XM_017022675.1 | XP_016878164.1 | 5 - 6 | 1957 | 101 | |
GenBank mRNA | AB058677.1 | - | 9 - 10 | 1346 | 101 |
AB300051.1 | - | 10 - 11 | 1412 | 101 | |
AL834326.1 | - | 4 - 5 | 667 | 101 | |
BC029999.1 | - | 11 - 12 | 1585 | 101 | |
BC117419.1 | - | 11 - 12 | 1473 | 101 | |
BC126313.1 | - | 11 - 12 | 1473 | 101 |