The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032445.2 | NP_115821.2 | 12 - 13 | 1717 | 60 |
XM_017022670.1 | XP_016878159.1 | 11 - 12 | 2139 | 60 | |
XM_017022671.1 | XP_016878160.1 | 11 - 12 | 2137 | 60 | |
XM_017022672.1 | XP_016878161.1 | 11 - 12 | 2146 | 60 | |
XM_017022673.1 | XP_016878162.1 | 11 - 12 | 2140 | 60 | |
XM_017022674.1 | XP_016878163.1 | 6 - 7 | 2083 | 60 | |
XM_017022675.1 | XP_016878164.1 | 6 - 7 | 2121 | 60 | |
GenBank mRNA | AB058677.1 | - | 10 - 11 | 1510 | 60 |
AB300051.1 | - | 11 - 12 | 1576 | 60 | |
AL834326.1 | - | 5 - 6 | 831 | 60 | |
BC029999.1 | - | 12 - 13 | 1749 | 60 | |
BC117419.1 | - | 12 - 13 | 1637 | 60 | |
BC126313.1 | - | 12 - 13 | 1637 | 60 |