The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286725.1 | NP_001273654.1 | 6 - 7 | 2245 | 70 |
NM_020866.2 | NP_065917.1 | 7 - 8 | 2428 | 70 | |
XM_017020678.1 | XP_016876167.1 | 7 - 8 | 1348 | 70 | |
XM_017020679.1 | XP_016876168.1 | 7 - 8 | 1935 | 70 | |
GenBank mRNA | AB040923.1 | - | 7 - 8 | 2394 | 70 |
AF252283.1 | - | 7 - 8 | 2262 | 70 | |
AK291435.1 | - | 7 - 8 | 2428 | 70 | |
AK295939.1 | - | 6 - 7 | 1436 | 70 | |
BC022460.1 | - | 7 - 8 | 1672 | 70 |