The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286725.1 | NP_001273654.1 | 7 - 8 | 2420 | 123 |
NM_020866.2 | NP_065917.1 | 8 - 9 | 2603 | 123 | |
XM_017020678.1 | XP_016876167.1 | 8 - 9 | 1523 | 123 | |
XM_017020679.1 | XP_016876168.1 | 8 - 9 | 2110 | 123 | |
GenBank mRNA | AB040923.1 | - | 8 - 9 | 2569 | 123 |
AF252283.1 | - | 8 - 9 | 2437 | 123 | |
AK291435.1 | - | 8 - 9 | 2603 | 123 | |
AK295939.1 | - | 7 - 8 | 1611 | 123 | |
BC022460.1 | - | 8 - 9 | 1847 | 123 |