The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199140.1 | NP_001186069.1 | 4 - 5 | 802 | 100 |
NM_031445.2 | NP_113633.2 | 4 - 5 | 774 | 100 | |
XM_005263804.4 | XP_005263861.1 | 4 - 5 | 1270 | 100 | |
XM_011511952.2 | XP_011510254.1 | 3 - 4 | 1160 | 100 | |
XM_011511955.2 | XP_011510257.1 | 2 - 3 | 189 | 100 | |
GenBank mRNA | AK304145.1 | - | 4 - 5 | 802 | 100 |
BC016181.1 | - | 4 - 5 | 774 | 100 | |
BC078174.1 | - | 4 - 5 | 784 | 100 |