Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP2
Entrez Gene ID
1844
Gene Name
dual specificity phosphatase 2
Gene Aliases
PAC-1, PAC1
Chromosome Location
Chr.2: 96143166 - 96145468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004418.3 NP_004409.1 1 - 2 479 57
GenBank mRNA BC007771.2 - 1 - 2 455 57
L11329.1 - 1 - 2 478 57

View Full Product Details