The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052934.3 | NP_443166.1 | 11 - 12 | 1410 | 103 |
NM_134325.2 | NP_599152.2 | 11 - 12 | 1410 | 103 | |
XM_011509121.2 | XP_011507423.1 | 10 - 11 | 1273 | 103 | |
XM_011509122.2 | XP_011507424.1 | 8 - 9 | 962 | 103 | |
XM_011509124.2 | XP_011507426.1 | 11 - 12 | 1422 | 103 | |
GenBank mRNA | AF314958.1 | - | 10 - 11 | 1298 | 103 |
AF331525.1 | - | 11 - 12 | 1410 | 103 | |
AK127491.1 | - | 3 - 4 | 680 | 103 | |
BC136538.1 | - | 11 - 12 | 1390 | 103 | |
BC144233.1 | - | 10 - 11 | 1279 | 103 | |
BC151208.1 | - | 11 - 12 | 1369 | 103 |