The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052934.3 | NP_443166.1 | 14 - 15 | 1679 | 125 |
NM_134325.2 | NP_599152.2 | 14 - 15 | 1679 | 125 | |
XM_011509121.2 | XP_011507423.1 | 13 - 14 | 1542 | 125 | |
XM_011509122.2 | XP_011507424.1 | 11 - 12 | 1231 | 125 | |
GenBank mRNA | AF314958.1 | - | 13 - 14 | 1567 | 125 |
AF331525.1 | - | 14 - 15 | 1679 | 125 | |
AK127491.1 | - | 6 - 7 | 949 | 125 | |
BC136538.1 | - | 14 - 15 | 1659 | 125 | |
BC144233.1 | - | 13 - 14 | 1507 | 125 | |
BC151208.1 | - | 14 - 15 | 1638 | 125 |