The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052934.3 | NP_443166.1 | 16 - 17 | 1885 | 163 |
NM_134325.2 | NP_599152.2 | 16 - 17 | 1885 | 163 | |
XM_011509121.2 | XP_011507423.1 | 15 - 16 | 1748 | 163 | |
XM_011509122.2 | XP_011507424.1 | 13 - 14 | 1437 | 163 | |
GenBank mRNA | AF314958.1 | - | 15 - 16 | 1773 | 163 |
AF331525.1 | - | 16 - 17 | 1885 | 163 | |
AK127491.1 | - | 8 - 9 | 1155 | 163 | |
BC136538.1 | - | 16 - 17 | 1865 | 163 | |
BC144233.1 | - | 15 - 16 | 1713 | 163 | |
BC151208.1 | - | 16 - 17 | 1844 | 163 |