Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHNAK2
Entrez Gene ID
113146
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
C14orf78
Chromosome Location
Chr.14: 104937253 - 104978357 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138420.2 NP_612429.2 7 - 7 1902 84
XM_005267299.1 XP_005267356.1 7 - 7 1755 84
GenBank mRNA AK094143.1 - 7 - 7 1755 84
BC004283.1 - 7 - 7 1754 84

View Full Product Details