The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003968.3 | NP_003959.3 | 7 - 8 | 490 | 140 |
NM_198195.1 | NP_937838.1 | 6 - 7 | 448 | 140 | |
XM_011534210.1 | XP_011532512.1 | 6 - 7 | 404 | 140 | |
XM_011534211.1 | XP_011532513.1 | 5 - 6 | 357 | 140 | |
GenBank mRNA | AB012190.1 | - | 5 - 6 | 407 | 140 |
AF046024.1 | - | 6 - 7 | 491 | 140 | |
AK002159.1 | - | 6 - 7 | 435 | 140 | |
AK289392.1 | - | 7 - 8 | 476 | 140 | |
AK314751.1 | - | 7 - 8 | 473 | 140 | |
AL117566.1 | - | 7 - 8 | 486 | 140 | |
BC022853.1 | - | 7 - 8 | 489 | 140 | |
CR533537.1 | - | 7 - 8 | 470 | 140 |