The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135651.2 | NP_001129123.1 | 3 - 4 | 435 | 138 |
NM_001135652.2 | NP_001129124.1 | 1 - 2 | 128 | 138 | |
NM_002759.3 | NP_002750.1 | 3 - 4 | 669 | 138 | |
XM_011532987.2 | XP_011531289.1 | 3 - 4 | 369 | 138 | |
XM_017004503.1 | XP_016859992.1 | 3 - 4 | 365 | 138 | |
GenBank mRNA | AK290655.1 | - | 3 - 4 | 355 | 138 |
AK313818.1 | - | 2 - 3 | 192 | 138 | |
AY302136.1 | - | 1 - 2 | 112 | 138 | |
BC007769.2 | - | 2 - 3 | 211 | 138 | |
BC022314.1 | - | 3 - 4 | 323 | 138 | |
BC040851.1 | - | 3 - 4 | 435 | 138 | |
BC057805.1 | - | 3 - 4 | 340 | 138 | |
BC093676.1 | - | 1 - 2 | 140 | 138 | |
BC101475.1 | - | 1 - 2 | 140 | 138 | |
BC143228.1 | - | 1 - 2 | 139 | 138 | |
BC143230.1 | - | 1 - 2 | 140 | 138 | |
M35663.1 | - | 2 - 3 | 298 | 138 | |
M85294.1 | - | 1 - 2 | 142 | 138 |