The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098520.1 | NP_001091990.1 | 2 - 3 | 368 | 76 |
NM_001098521.1 | NP_001091991.1 | 2 - 3 | 296 | 76 | |
NM_001098522.1 | NP_001091992.1 | 1 - 2 | 578 | 76 | |
NM_001098523.1 | NP_001091993.1 | 1 - 2 | 294 | 76 | |
NM_006410.4 | NP_006401.3 | 2 - 3 | 263 | 76 | |
GenBank mRNA | AF039103.1 | - | 1 - 2 | 209 | 76 |
AF092095.1 | - | 1 - 2 | 295 | 76 | |
AK222969.1 | - | 1 - 2 | 252 | 76 | |
AK223010.1 | - | 1 - 2 | 286 | 76 | |
AK292092.1 | - | 2 - 3 | 272 | 76 | |
AW963626.1 | - | 1 - 2 | 213 | 76 | |
BC002439.2 | - | 2 - 3 | 309 | 76 | |
BC015358.2 | - | 1 - 2 | 274 | 76 | |
BF681178.1 | - | - | 262 | 76 | |
BP382529.1 | - | 2 - 3 | 368 | 76 | |
CR456821.1 | - | 1 - 2 | 197 | 76 | |
DB450790.1 | - | 2 - 3 | 295 | 76 | |
U69161.1 | - | 1 - 2 | 295 | 76 |