The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278716.1 | NP_001265645.1 | 8 - 9 | 1893 | 75 |
NM_012160.4 | NP_036292.2 | 7 - 8 | 1775 | 75 | |
NR_103836.1 | 6 - 7 | 1429 | 75 | ||
XM_017010726.1 | XP_016866215.1 | 7 - 8 | 1707 | 75 | |
XM_017010728.1 | XP_016866217.1 | 9 - 10 | 1969 | 75 | |
GenBank mRNA | AF174590.1 | - | 5 - 6 | 1384 | 75 |
AF176699.1 | - | 5 - 6 | 1384 | 75 | |
AF199355.1 | - | 5 - 6 | 1388 | 75 | |
AK313076.1 | - | 7 - 8 | 1704 | 75 | |
BC055010.1 | - | 7 - 8 | 1714 | 75 | |
BC091484.1 | - | 8 - 9 | 1813 | 75 |