Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCRB1
Entrez Gene ID
85437
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Gene Aliases
MADP-1, MADP1, RBM36, SNRNP31, ZCCHC19
Chromosome Location
Chr.12: 42312086 - 42326130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033114.3 NP_149105.3 4 - 5 431 88
XM_017020124.1 XP_016875613.1 3 - 4 322 88
GenBank mRNA AB062361.1 - 4 - 5 431 88
AK308244.1 - 4 - 5 301 88
BC005099.1 - 4 - 5 381 88
BC010177.2 - 4 - 5 396 88
BC022543.1 - 4 - 5 406 88

View Full Product Details