The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282723.1 | NP_001269652.1 | 4 - 5 | 569 | 95 |
NM_001282724.1 | NP_001269653.1 | 3 - 4 | 443 | 95 | |
NR_104233.1 | 3 - 4 | 413 | 95 | ||
GenBank mRNA | AK299244.1 | - | 4 - 5 | 510 | 95 |
AK300494.1 | - | 3 - 4 | 365 | 95 | |
AK301493.1 | - | 3 - 4 | 414 | 95 |