The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014009.3 | NP_054728.2 | 2 - 3 | 403 | 65 |
XM_006724533.2 | XP_006724596.2 | 2 - 3 | 509 | 65 | |
GenBank mRNA | AF277993.1 | - | 2 - 3 | 403 | 65 |
AK292052.1 | - | 2 - 3 | 287 | 65 | |
BC113401.1 | - | 1 - 2 | 245 | 65 | |
BC113403.1 | - | 1 - 2 | 245 | 65 | |
BC143786.1 | - | 1 - 2 | 245 | 65 | |
EF534714.1 | - | 1 - 2 | 215 | 65 | |
EU855812.1 | - | 1 - 2 | 215 | 65 |