The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258462.1 | NP_001245391.1 | 6 - 7 | 655 | 114 |
NM_001258463.1 | NP_001245392.1 | 6 - 7 | 593 | 114 | |
NM_001310158.1 | NP_001297087.1 | 6 - 7 | 696 | 114 | |
NM_001310160.1 | NP_001297089.1 | 2 - 3 | 1579 | 114 | |
NM_001310161.1 | NP_001297090.1 | 3 - 4 | 462 | 114 | |
NM_001604.5 | NP_001595.2 | 6 - 7 | 643 | 114 | |
GenBank mRNA | AB593092.1 | - | 6 - 7 | 594 | 114 |
AB593093.1 | - | 6 - 7 | 593 | 114 | |
AB593094.1 | - | 6 - 7 | 696 | 114 | |
AK094249.1 | - | 2 - 3 | 1389 | 114 | |
BX640762.1 | - | 6 - 7 | 622 | 114 | |
DA089215.1 | - | 3 - 4 | 462 | 114 | |
DA571138.1 | - | 3 - 4 | 284 | 114 | |
KP255960.1 | - | 2 - 3 | 186 | 114 |