The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286075.1 | NP_001273004.1 | 10 - 11 | 1254 | 120 |
NM_004320.4 | NP_004311.1 | 12 - 13 | 1605 | 120 | |
NM_173201.3 | NP_775293.1 | 12 - 13 | 1605 | 120 | |
GenBank mRNA | AK128456.1 | - | 10 - 11 | 1244 | 120 |
AK291314.1 | - | 12 - 13 | 1534 | 120 | |
BC037354.1 | - | 12 - 13 | 1533 | 120 | |
BX537389.1 | - | 12 - 13 | 1601 | 120 | |
BX537784.1 | - | 12 - 13 | 1603 | 120 | |
BX647365.1 | - | 12 - 13 | 1601 | 120 | |
BX647366.1 | - | 12 - 13 | 1597 | 120 | |
BX647367.1 | - | 12 - 13 | 1605 | 120 | |
BX647368.1 | - | 5 - 6 | 643 | 120 | |
BX648544.1 | - | 12 - 13 | 1602 | 120 | |
BX649072.1 | - | 12 - 13 | 1593 | 120 |