The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286075.1 | NP_001273004.1 | 11 - 12 | 1374 | 68 |
NM_004320.4 | NP_004311.1 | 13 - 14 | 1725 | 68 | |
NM_173201.3 | NP_775293.1 | 13 - 14 | 1725 | 68 | |
GenBank mRNA | AK128456.1 | - | 11 - 12 | 1364 | 68 |
AK291314.1 | - | 13 - 14 | 1654 | 68 | |
BC037354.1 | - | 13 - 14 | 1653 | 68 | |
BX537389.1 | - | 13 - 14 | 1721 | 68 | |
BX537784.1 | - | 13 - 14 | 1723 | 68 | |
BX647365.1 | - | 13 - 14 | 1721 | 68 | |
BX647366.1 | - | 13 - 14 | 1717 | 68 | |
BX647367.1 | - | 13 - 14 | 1725 | 68 | |
BX647368.1 | - | 6 - 7 | 763 | 68 | |
BX649072.1 | - | 13 - 14 | 1713 | 68 |