Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP82
Entrez Gene ID
284406
Gene Name
ZFP82 zinc finger protein
Gene Aliases
ZNF545
Chromosome Location
Chr.19: 36383102 - 36418674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321917.1 NP_001308846.1 1 - 2 189 96
NM_001321918.1 NP_001308847.1 1 - 2 189 96
NM_001321919.1 NP_001308848.1 1 - 2 189 96
NM_133466.3 NP_597723.1 1 - 2 189 96
GenBank mRNA AK074942.1 - 1 - 2 171 96
AK315709.1 - 1 - 2 149 96
AL834267.1 - 1 - 2 163 96
BC032577.2 - 1 - 2 156 96
BX491216.1 - 1 - 2 190 96
GU388429.1 - 1 - 2 159 96

View Full Product Details