Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCK
Entrez Gene ID
885
Gene Name
cholecystokinin
Gene Aliases
-
Chromosome Location
Chr.3: 42257824 - 42266195 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000729.5 NP_000720.1 2 - 3 681 58
GenBank mRNA BC111026.1 - 2 - 3 656 58
BM544754.1 - 2 - 3 704 58

View Full Product Details