Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
Chromosome Location
Chr.9: 120798318 - 120843021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270427.1 NP_001257356.1 2 - 3 434 152
NM_005047.3 NP_005038.1 2 - 3 434 152
XM_011518866.2 XP_011517168.1 2 - 3 344 152
XM_017014927.1 XP_016870416.1 2 - 3 344 152
XM_017014928.1 XP_016870417.1 2 - 3 344 152
GenBank mRNA AK291051.1 - 2 - 3 335 152
AK303007.1 - 2 - 3 364 152
BC014478.1 - 2 - 3 323 152
BM835267.1 - 2 - 3 341 152
D31889.1 - 2 - 3 321 152
S79862.1 - 2 - 3 325 152

View Full Product Details